Imprimer

Anti-Influenza A M1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Biorbyt
BIRBORB23147-1EA 635 CHF
BIRBORB23147-1
Anti-Influenza A M1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Type: Primary
Antigen: Influenza A M1
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR