Imprimer

Anti-Neisseria gonorrhoeae Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Biorbyt
BIRBORB23091-1EA 656 CHF
BIRBORB23091-1
Anti-Neisseria gonorrhoeae Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Type: Primary
Antigen: Neisseria gonorrhoeae
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR