Imprimer

Anti-Rotavirus Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB23088-1EA 710 CHF
BIRBORB23088-1
Anti-Rotavirus Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Rotavirus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR