Imprimer

Anti-EBV Nuclear Antigen Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB23081-1EA 581 CHF
BIRBORB23081-1
Anti-EBV Nuclear Antigen Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: EBV Nuclear Antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR