Imprimer
Anti-L. pneumophila Rabbit Polyclonal Antibody
Fournisseur: Biorbyt
BIRBORB23073-1EA 0 CHF
BIRBORB23073-1
Anti-L. pneumophila Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: L. pneumophila
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now