Imprimer

Anti-MAPK3 Mouse Monoclonal Antibody [clone: E19]

Fournisseur: Biorbyt
BIRBORB22970-100EA 0 CHF
BIRBORB22970-100
Anti-MAPK3 Mouse Monoclonal Antibody [clone: E19]
Anticorps
Type: Primary
Antigen: MAPK3 (mitogen-activated protein kinase 3)
Clonality: monoclonal
Clone: E19
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR