Imprimer

Anti-ACTB Mouse Monoclonal Antibody [clone: BA3R]

Fournisseur: Biorbyt
BIRBORB22968-100EA 0 CHF
BIRBORB22968-100
Anti-ACTB Mouse Monoclonal Antibody [clone: BA3R]
Anticorps
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: monoclonal
Clone: BA3R
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR