Imprimer

Anti-TGFBI Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB22912-5EA 0 CHF
BIRBORB22912-5 BIRBORB22912-100 BIRBORB22912-20
Anti-TGFBI Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TGFBI (transforming growth factor, beta-induced, 68kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR