Imprimer
Anti-Streptavidin Mouse Monoclonal Antibody
Fournisseur: Biorbyt
BIRBORB22909-100EA 789 CHF
BIRBORB22909-100 BIRBORB22909-20 BIRBORB22909-5
Anti-Streptavidin Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Streptavidin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity:
Order Now