Imprimer

Anti-RASSF1A Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB22905-20EA 404 CHF
BIRBORB22905-20 BIRBORB22905-100 BIRBORB22905-5
Anti-RASSF1A Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RASSF1A
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR