Imprimer

Anti-MRSA Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB22885-500EA 613 CHF
BIRBORB22885-500 BIRBORB22885-1
Anti-MRSA Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: MRSA
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR