Imprimer

Anti-MHC Class II (I-E) Mouse Monoclonal Antibody (FITC (Fluorescein))

Fournisseur: Biorbyt
BIRBORB22863-1EA 0 CHF
BIRBORB22863-1 BIRBORB22863-500
Anti-MHC Class II (I-E) Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorps
Type: Primary
Antigen: MHC Class II (I-E)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR