Imprimer

Anti-MHC Class II Mouse Monoclonal Antibody (Biotin)

Fournisseur: Biorbyt
BIRBORB22859-1EA 0 CHF
BIRBORB22859-1 BIRBORB22859-500
Anti-MHC Class II Mouse Monoclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: MHC Class II
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR