Imprimer
Alcool absolu 94.0-96.0%, AR® ACS (denaturé avec 5% Propanol-2 et 5% methanol), Macron Fine Chemicals™
7019-28EA 0 CHF
7019-02 MACR7019-10 7019-28
Alcool absolu 94.0-96.0%, AR® ACS (denaturé avec 5% Propanol-2 et 5% methanol), Macron Fine Chemicals™
Alcohol
température de stockage: Ambient
MDL Number: MFCD00003568
Numéro CAS: 64-17-5
ONU: 1987
ADR: 3,II
Order Now
Résultats des tests de spécification
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Specially Denatured Alcohol Formula 3A (200 proof)(v/v) 94.0 - 96.0 %
Ethanol (v/v) 88.5 - 92.5 %
Methanol (v/v) 3.5 - 5.5 %
Isopropyl Alcohol (2-propanol)(v/v) 4.0 - 6.0 %
Total Alcohol (v/v) ≥98.0 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Specific Gravity at 25°C 0.782 - 0.793
Water (H₂O) ≤0.5 %
SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200
proof ethanol. Product Information (not specifications): Appearance
(clear, colorless liquid)