Acide 2-bromo-3-nitrobenzoïque ≥90%, tech.
Fournisseur: Thermo Scientific
|
Attention
|
H60795.03EA
128
CHF
H60795.03
Acide 2-bromo-3-nitrobenzoïque ≥90%, tech.
Acide 2-bromo-3-nitrobenzoïque
Formule:
C₇H₄BrNO₄ MW: 246,02 g/mol Melting Pt: 184…186 °C température de stockage: Température ambiante |
MDL Number:
MFCD00074899 Numéro CAS: 573-54-6 EINECS: 209-356-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...