Imprimer

1-Cyclopentene-1-carboxylic acid

Fournisseur: ACTIVATE SCIENTIFIC GMBH

ACTIAS35291G1EA 0 CHF
ACTIAS35291G1 ACTIAS35291G10 ACTIAS35291G5
1-Cyclopentene-1-carboxylic acid
1-Cyclopentene-1-carboxylic acid
Formule: C₆H₈O₂
MW: 112,13 g/mol
MDL Number: MFCD00001396
Numéro CAS: 1560-11-8
ONU: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR