Imprimer

3-(4-Hydroxyphényl)-1-(2,4,6-trihydroxyphényl)propane-1-one

Fournisseur: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-(4-Hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)propan-1-one

ACTIAS24974G5EA 0 CHF
ACTIAS24974G5 ACTIAS24974G25
3-(4-Hydroxyphényl)-1-(2,4,6-trihydroxyphényl)propane-1-one
3-(4-Hydroxyphényl)-1-(2,4,6-trihydroxyphényl)propane-1-one
Formule: C₁₅H₁₄O₅
MW: 274,28 g/mol
MDL Number: MFCD00002288
Numéro CAS: 60-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR