2-Amino-4-chlorobenzamide
Fournisseur: ACTIVATE SCIENTIFIC GMBH
ACTIAS36333G1EA
36
CHF
ACTIAS36333MG250
ACTIAS36333G1
ACTIAS36333G5
2-Amino-4-chlorobenzamide
2-Amino-4-chlorobenzamide
Formule:
C₇H₇ClN₂O MW: 170,6000 g/mol |
MDL Number:
MFCD09042792 Numéro CAS: 5900-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...