2-Chloro-3-méthylquinoléine
Fournisseur: ACTIVATE SCIENTIFIC GMBH
ACTIAS35282G1EA
0
CHF
ACTIAS35282G1
ACTIAS35282G10
ACTIAS35282G5
2-Chloro-3-méthylquinoléine
2-Chloro-3-méthylquinoléine
Formule:
C₁₀H₈ClN MW: 177,63 g/mol |
MDL Number:
MFCD00100265 Numéro CAS: 57876-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...