α,α,α-Trifluoro-2-tolualdehyde
Fournisseur: ACTIVATE SCIENTIFIC GMBH
Synonyms:
2-(Trifluorométhyl)benzaldéhyde
ACTIAS40093G100EA
0
CHF
ACTIAS40093G100
α,α,α-Trifluoro-2-tolualdehyde
α,α,α-Trifluoro-2-tolualdehyde
Formule:
C₈H₅F₃O MW: 174,12 g/mol Point d'ébullition: 28…38 °C (0,4 mmHg) Melting Pt: –40 °C Densité: 1,322 g/cm³ (20 °C) |
MDL Number:
MFCD00003337 Numéro CAS: 447-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...