4-Fluoroindole
Fournisseur: ACTIVATE SCIENTIFIC GMBH
ACTIAS50516G5EA
0
CHF
ACTIAS50516G5
ACTIAS50516G25
4-Fluoroindole
4-Fluoroindole
Formule:
C₈H₆FN MW: 135,14 g/mol |
MDL Number:
MFCD00055992 Numéro CAS: 387-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...