Imprimer

2-Fluoro-3-méthylpyridine

Fournisseur: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Fluoro-3-picoline

ACTIAS1376G5EA 0 CHF
ACTIAS1376G5
2-Fluoro-3-méthylpyridine
2-Fluoro-3-méthylpyridine
Formule: C₆H₆FN
MW: 111,12 g/mol
MDL Number: MFCD00041223
Numéro CAS: 2369-18-8
ONU: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR