Imprimer

Acide 4-hydroxy-3,5-diméthoxycinnamique

Fournisseur: ACTIVATE SCIENTIFIC GMBH

ACTIAS25293G100EA 0 CHF
ACTIAS25293G100 ACTIAS25293G5 ACTIAS25293G25
Acide 4-hydroxy-3,5-diméthoxycinnamique
Acide 4-hydroxy-3,5-diméthoxycinnamique
Formule: C₁₁H₁₂O₅
MW: 224,21 g/mol
Point d'ébullition: 378 °C (1013 hPa)
Melting Pt: 200…202 °C
MDL Number: MFCD00004401
Numéro CAS: 530-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR