3,6-Dihydroxyphtalonitrile
Fournisseur: ACTIVATE SCIENTIFIC GMBH
ACTIAS23865G100EA
205
CHF
ACTIAS23865G100
3,6-Dihydroxyphtalonitrile
3,6-Dihydroxyphtalonitrile
Formule:
C₈H₄N₂O₂ MW: 160,13 g/mol |
MDL Number:
MFCD00001790 Numéro CAS: 4733-50-0 ONU: 3439 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...