2,4-Dichloro-5-nitropyrimidine
Fournisseur: ACTIVATE SCIENTIFIC GMBH
ACTIAS7175G5EA
0
CHF
ACTIAS7175G50
ACTIAS7175G5
2,4-Dichloro-5-nitropyrimidine
2,4-Dichloro-5-nitropyrimidine
Formule:
C₄HCl₂N₃O₂ MW: 193,98 g/mol Point d'ébullition: 326 °C (1013 hPa) Melting Pt: 29,3 °C Densité: 1,74 g/cm³ (20 °C) |
MDL Number:
MFCD00127867 Numéro CAS: 49845-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...