Imprimer

2,4-Dicloro-6-(trifluorométhyl)aniline

Fournisseur: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4,6-Dichloro-α,α,α-trifluoro-o-toluidine

ACTIAS25370G25EA 290 CHF
ACTIAS25370G25
2,4-Dicloro-6-(trifluorométhyl)aniline
2,4-Dicloro-6-(trifluorométhyl)aniline
Formule: C₇H₄Cl₂F₃N
MW: 230,02 g/mol
MDL Number: MFCD00174313
Numéro CAS: 62593-17-3
ONU: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR