2-Chloro-5-fluorobenzonitrile
Fournisseur: ACTIVATE SCIENTIFIC GMBH
ACTIAS34917G10EA
0
CHF
ACTIAS34917G10
ACTIAS34917G5
ACTIAS34917G1
2-Chloro-5-fluorobenzonitrile
2-Chloro-5-fluorobenzonitrile
Formule:
C₇H₃ClFN MW: 155,56 g/mol |
MDL Number:
MFCD03094168 Numéro CAS: 57381-56-3 ONU: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...