Acide 2-chloro-6-méthoxybenzoïque
Fournisseur: ACTIVATE SCIENTIFIC GMBH
ACTIAS30832G5EA
0
CHF
ACTIAS30832G5
ACTIAS30832G1
Acide 2-chloro-6-méthoxybenzoïque
Acide 2-chloro-6-méthoxybenzoïque
Formule:
C₈H₇ClO₃ |
MDL Number:
MFCD00127702 Numéro CAS: 3260-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...