2-Bromo-4-iodopyridine
Fournisseur: ACTIVATE SCIENTIFIC GMBH
ACTIAS31690G5EA
0
CHF
ACTIAS31690G5
ACTIAS31690G1
2-Bromo-4-iodopyridine
2-Bromo-4-iodopyridine
Formule:
C₅H₃BrIN MW: 283,89 g/mol |
MDL Number:
MFCD03085768 Numéro CAS: 100523-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...