Imprimer

4-Bromo-2-chloro-α,α,α-trifluorotoluène

Fournisseur: ACTIVATE SCIENTIFIC GMBH

ACTIAS33358G5EA 0 CHF
ACTIAS33358G10 ACTIAS33358G5
4-Bromo-2-chloro-α,α,α-trifluorotoluène
4-Bromo-2-chloro-α,α,α-trifluorotoluène
Formule: C₇H₃BrClF₃
MW: 259,45 g/mol
MDL Number: MFCD08459292
Numéro CAS: 467435-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR