Acide (2-bromophényl)métaborique
Fournisseur: ACTIVATE SCIENTIFIC GMBH
Synonyms:
Dihydroxy-2-bromophénylborane
ACTIAS32327G10EA
0
CHF
ACTIAS32327G10
ACTIAS32327G5
ACTIAS32327G1
Acide (2-bromophényl)métaborique
Acide (2-bromophényl)métaborique
Formule:
C₆H₆BBrO₂ MW: 200,83 g/mol |
MDL Number:
MFCD01114672 Numéro CAS: 244205-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...