2,4-Dibromopyridine
Fournisseur: ACTIVATE SCIENTIFIC GMBH
ACTIAS50201G5EA
0
CHF
ACTIAS50201G25
ACTIAS50201G1
ACTIAS50201G5
2,4-Dibromopyridine
2,4-Dibromopyridine
Formule:
C₅H₃Br₂N MW: 236,89 g/mol |
MDL Number:
MFCD01859720 Numéro CAS: 58530-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...