Gas inlet tubes, BOLA
Supplier: Bohlender
BOLA
BOHLN1502-06EA 34.2 CHF
BOHLN1502-06 BOHLN1502-04 BOHLN1502-02
Gas inlet tubes, BOLA
Tubes Gas Inlet Tubes
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
For constructing a gas washing equipment. Tube with Int.Ø 5 mm, one side with hose connector dia. 9 mm for connecting tubing, other side with thread M 8×1 for connecting a gas frit or a gas distributor.
Mehr About VWR