2-Bromo-5-fluoro-N-isobutylbenzamide

Supplier: Apollo Scientific

APOSPC8014-1GEA 211 CHF
APOSPC8014-1G
2-Bromo-5-fluoro-N-isobutylbenzamide
2-Bromo-5-fluoro-N-isobutylbenzamide
Formula: C₁₁H₁₃BrFNO
Storage Temperature: Ambient
MDL Number: MFCD09800914
CAS Number: 951884-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR