3-(4-Ethoxy-3-(trifluoromethyl)phenyl)propionic acid 98%

Supplier: Apollo Scientific

APOSPC302769-1GEA 245 CHF
APOSPC302769-1G APOSPC302769-5G
3-(4-Ethoxy-3-(trifluoromethyl)phenyl)propionic acid 98%
3-(4-Ethoxy-3-(trifluoromethyl)phenyl)propionic acid
Formula: C₁₂H₁₃F₃O₃
MW: 262.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13196589
CAS Number: 1206593-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR