3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride

Supplier: Apollo Scientific
Warning

APOSPC2714-25GEA 69.8 CHF
APOSPC2714-25G APOSPC2714-5G APOSPC2714-1G
3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride
3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride
Formula: C₈H₇ClF₆N₂
MW: 280.60 g/mol
Melting Pt: ca 208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03094165
CAS Number: 502496-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR