Pentafluorophenyl-5-bromo-2-thiophenesulphonate

Supplier: Apollo Scientific

APOSPC10565-25GEA 1410 CHF
APOSPC10565-25G APOSPC10565-1G APOSPC10565-5G
Pentafluorophenyl-5-bromo-2-thiophenesulphonate
Pentafluorophenyl 5-bromo-2-thiophenesulphonate
Formula: C₁₀H₂BrF₅O₃S₂
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD06659553
CAS Number: 885949-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR