Pentafluorophenyl-2-nitrobenzenesulphonate

Supplier: Apollo Scientific

APOSPC10319-1GEA 330 CHF
APOSPC10319-1G APOSPC10319-500MG APOSPC10319-5G
Pentafluorophenyl-2-nitrobenzenesulphonate
Pentafluorophenyl 2-nitrobenzenesulphonate
Formula: C₁₂H₄F₅NO₅S
Melting Pt: 127…128 °C
Storage Temperature: Ambient
MDL Number: MFCD05975122
CAS Number: 885950-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR