4-Bromo-3-nitrobenzoic acid

Supplier: Apollo Scientific
Warning

APOSOR8176-100GEA 300 CHF
APOSOR8176-100G APOSOR8176-5G APOSOR8176-25G
4-Bromo-3-nitrobenzoic acid
4-Bromo-3-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 199…206 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00272137
CAS Number: 6319-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR