2-Chloro-5-iodophenol

Supplier: Apollo Scientific
Danger

APOSOR6603-1GEA 17.6 CHF
APOSOR6603-1G APOSOR6603-5G APOSOR6603-25G
2-Chloro-5-iodophenol
2-Chloro-5-iodophenol
Formula: C₆H₄ClIO
MW: 254.4500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672969
CAS Number: 289039-26-5
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR