4-Bromobenzyl alcohol 98%

Supplier: Apollo Scientific
Warning

APOSOR5021-25GEA 17.6 CHF
APOSOR5021-25G APOSOR5021-100G APOSOR5021-500G
4-Bromobenzyl alcohol 98%
4-Bromobenzyl alcohol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00004650
CAS Number: 873-75-6
EINECS: 212-851-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR