1,2-Dimethoxy-4,5-dinitrobenzene

Supplier: Apollo Scientific
Warning

APOSOR28313-10GEA 392 CHF
APOSOR28313-10G APOSOR28313-1G APOSOR28313-5G
1,2-Dimethoxy-4,5-dinitrobenzene
1,2-Dimethoxy-4,5-dinitrobenzene
Formula: C₈H₈N₂O₆
MW: 228.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082695
CAS Number: 3395-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR