4-(2-Carboxyethyl)phenylboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 4-(2-Carboxyethyl)benzeneboronic acid

APOSOR13109-1GEA 114 CHF
APOSOR13109-1G APOSOR13109-5G
4-(2-Carboxyethyl)phenylboronic acid
4-(2-Carboxyethyl)phenylboronic acid
Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318119
CAS Number: 166316-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR