2-Chloro-4'-methylacetanilide

Supplier: Apollo Scientific

Synonyms: N-(Chloroacetyl)-4-methylaniline

APOSOR12180-1GEA 0 CHF
APOSOR12180-1G
2-Chloro-4'-methylacetanilide
2-Chloro-4'-methylacetanilide
Formula: C₉H₁₀ClNO
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00018910
CAS Number: 16634-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR