Ethyl-2-(dimethylamino)benzoate 97%

Supplier: Apollo Scientific

APOSOR963928-25GEA 908 CHF
APOSOR963928-25G APOSOR963928-1G APOSOR963928-5G
Ethyl-2-(dimethylamino)benzoate 97%
Ethyl 2-(dimethylamino)benzoate
Formula: C₁₁H₁₅NO₂
MW: 193.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009110
CAS Number: 55426-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR