1-(Cyclopropylmethyl)thiourea 97%

Supplier: Apollo Scientific
Warning

Synonyms: (Thioureidomethyl)cyclopropane[(Carbamothioylamino)methyl]cyclopropane

APOSOR28359-5GEA 0 CHF
APOSOR28359-5G APOSOR28359-250MG APOSOR28359-1G
1-(Cyclopropylmethyl)thiourea 97%
1-(Cyclopropylmethyl)thiourea
Formula: C₅H₁₀N₂S
MW: 130.211 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00276905
CAS Number: 618913-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR