3-Amino-4-(methylamino)benzonitrile

Supplier: Apollo Scientific
Warning

APOSOR25947-5GEA 0 CHF
APOSOR25947-5G APOSOR25947-25G APOSOR25947-1G
3-Amino-4-(methylamino)benzonitrile
3-Amino-4-(methylamino)benzonitrile
Formula: C₈H₉N₃
MW: 147.177 g/mol
Melting Pt: 136…140 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03407328
CAS Number: 64910-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR