2-(Morpholin-4-yl)-1-phenylethylamine 95+%

Supplier: Apollo Scientific
Danger

Synonyms: 4-(2-Amino-2-phenylethyl)morpholineα-[(Morpholin-4-yl)methyl]benzylamine

APOSOR12713-1GEA 399 CHF
APOSOR12713-1G APOSOR12713-100MG APOSOR12713-250MG
2-(Morpholin-4-yl)-1-phenylethylamine 95+%
2-(Morpholin-4-yl)-1-phenylethylamine
Formula: C₁₂H₁₈N₂O
MW: 206.284 g/mol
Boiling Pt: 130…134 °C (2 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01862532
CAS Number: 38060-08-1
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR