2-Amino-4-(ethoxycarbonyl)benzeneboronic acid hydrochloride 95%

Supplier: Apollo Scientific

Synonyms: Ethyl 3-amino-4-boronobenzoate hydrochloride

APOSOR11454-1GEA 798 CHF
APOSOR11454-1G
2-Amino-4-(ethoxycarbonyl)benzeneboronic acid hydrochloride 95%
2-Amino-4-(ethoxycarbonyl)benzeneboronic acid hydrochloride
Formula: C₉H₁₂BNO₄·ClH
MW: 245.468 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09475809
CAS Number: 957034-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR