2-Chloro-3-(pyrrolidin-1-ylsulphonyl)pyridine 97%

Supplier: Apollo Scientific

Synonyms: 1-[(2-Chloropyridin-3-yl)sulphonyl]pyrrolidine

APOSOR11432-250MGEA 0 CHF
APOSOR11432-250MG APOSOR11432-1G
2-Chloro-3-(pyrrolidin-1-ylsulphonyl)pyridine 97%
2-Chloro-3-(pyrrolidin-1-ylsulphonyl)pyridine
Formula: C₉H₁₁ClN₂O₂S
MW: 246.714 g/mol
Melting Pt: 62…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09475828
CAS Number: 60597-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR